Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DISP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033510.2 | NP_277045.1 | 8 | 4376 | ||
XM_011522125.2 | XP_011520427.1 | 9 | 6547 | |||
XM_017022687.1 | XP_016878176.1 | 9 | 6547 | |||
GenBank mRNA | AB051529.2 | - | 8 | 4381 | ||
AK096153.1 | - | 1 | 2059 | |||
CR627480.1 | - | 1 | 457 |