Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTNAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033655.3 | NP_387504.2 | 7 | 1223 | ||
GenBank mRNA | AB051501.2 | - | 7 | 1130 | ||
AF333769.2 | - | 7 | 1091 | |||
AK056833.1 | - | 7 | 1223 | |||
AK294281.1 | - | 7 | 1015 | |||
BC132737.1 | - | 7 | 1128 | 40146759 | ||
BC144130.1 | - | 7 | 1128 |
Gene Symbol: LOC100289279 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545672.2 | XP_011543974.1 | 7 | 1360 | ||
XM_011545673.2 | XP_011543975.1 | 7 | 1360 | |||
XM_011545674.2 | XP_011543976.1 | 7 | 1360 | |||
XM_017015380.1 | XP_016870869.1 | 7 | 1360 |