Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052813.4 | NP_434700.2 | 13 | 2100 | ||
NM_052814.3 | NP_434701.1 | 12 | 1791 | |||
GenBank mRNA | AF311287.1 | - | 13 | 2058 | ||
AK024001.1 | - | 14 | 2154 | |||
AK127481.1 | - | 5 | 4555 | |||
AK292081.1 | - | 13 | 2052 | |||
BC008877.2 | - | 12 | 1766 | 4139529 |