Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128855.2 | NP_001122327.1 | 5 | 706 | ||
NM_001195422.1 | NP_001182351.1 | 5 | 699 | |||
NM_032620.3 | NP_116009.2 | 5 | 706 | |||
NM_133644.3 | NP_598399.2 | 5 | 706 | |||
GenBank mRNA | AF360742.1 | - | 5 | 668 | ||
AF360743.1 | - | 5 | 668 | |||
AF360744.1 | - | 5 | 648 | |||
AK027606.1 | - | 5 | 653 | |||
AK091035.1 | - | 5 | 1411 | |||
AK291929.1 | - | 5 | 650 | |||
AK297953.1 | - | 5 | 681 | |||
AK298485.1 | - | 929 | ||||
AK311442.1 | - | 5 | 769 | |||
AY078987.1 | - | 5 | 668 | |||
AY078988.1 | - | 5 | 668 | |||
BC008924.2 | - | 3 | 246 | |||
BC017207.2 | - | 5 | 638 | 3634066 | ||
BC019261.1 | - | 5 | 647 | 3501739 | ||
BM789389.1 | - | 2 | 170 | |||
DA490131.1 | - | 1 | 214 |