Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202476.1 | NP_001189405.1 | 5 | 681 | ||
NM_003341.4 | NP_003332.1 | 6 | 976 | |||
NM_182666.2 | NP_872607.1 | 5 | 925 | |||
GenBank mRNA | BC009139.1 | - | 6 | 900 | 3853408 |