Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138376.2 | NP_612385.2 | 3 | 294 | ||
GenBank mRNA | AK074553.1 | - | 3 | 277 | ||
BC008647.1 | - | 3 | 235 | |||
BC030822.1 | - | 3 | 238 | 4243419 | ||
BC053538.1 | - | 3 | 242 | 5225152 | ||
BX248066.1 | - | 3 | 231 | |||
BX640732.1 | - | 3 | 241 | |||
BX648584.1 | - | 3 | 249 | |||
DB051711.1 | - | 3 | 294 |