Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173512.1 | NP_001166983.1 | 6 | 1792 | ||
NM_138777.3 | NP_620132.1 | 7 | 1948 | |||
NM_199177.2 | NP_954646.1 | 5 | 1452 | |||
XM_005252302.3 | XP_005252359.1 | 8 | 2016 | |||
XM_011519183.2 | XP_011517485.1 | 8 | 2271 | |||
XM_011519184.2 | XP_011517486.1 | 8 | 1781 | |||
XM_011519185.2 | XP_011517487.1 | 7 | 1856 | |||
XM_011519186.1 | XP_011517488.1 | 6 | 1536 | |||
XM_011519187.2 | XP_011517489.1 | 6 | 1736 | |||
XM_017015286.1 | XP_016870775.1 | 7 | 1924 | |||
XM_017015287.1 | XP_016870776.1 | 6 | 1604 | |||
GenBank mRNA | BC013049.1 | - | 6 | 1609 | 3882448 |