Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138961.2 | NP_620411.2 | 5 | 810 | ||
XM_005271720.4 | XP_005271777.1 | 2 | 231 | |||
XM_011543060.2 | XP_011541362.1 | 4 | 573 | |||
GenBank mRNA | AF361746.1 | - | 5 | 775 | ||
AK075396.1 | - | 5 | 773 | |||
AK091023.1 | - | 5 | 1693 | |||
AK092429.1 | - | 3 | 1182 | |||
AK301160.1 | - | 2 | 236 | |||
AK314710.1 | - | 5 | 773 | |||
AY189281.1 | - | 2 | 1356 | |||
AY358382.1 | - | 5 | 762 | |||
BC016868.1 | - | 5 | 644 | 3850665 |