Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138961.2 | NP_620411.2 | 7 | 1722 | ||
XM_005271720.4 | XP_005271777.1 | 4 | 1143 | |||
XM_011543060.2 | XP_011541362.1 | 6 | 1485 | |||
GenBank mRNA | AF277292.1 | - | 1 | 635 | ||
AF361746.1 | - | 7 | 1687 | |||
AK075396.1 | - | 7 | 1685 | |||
AK091023.1 | - | 8 | 2568 | |||
AK092429.1 | - | 5 | 2094 | |||
AY189281.1 | - | 4 | 2279 | |||
AY358382.1 | - | 7 | 1674 | |||
BC016868.1 | - | 7 | 1556 | 3850665 |