Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144570.2 | NP_653171.1 | 5 | 715 | ||
GenBank mRNA | AK023154.1 | - | 5 | 713 | ||
AK296888.1 | - | 4 | 411 | |||
AK296998.1 | - | 6 | 803 | |||
AK303927.1 | - | 5 | 620 | |||
AK304025.1 | - | 5 | 797 | |||
AK309909.1 | - | 4 | 587 | |||
AK309930.1 | - | 4 | 536 | |||
AK316514.1 | - | 5 | 771 | |||
AY323978.1 | - | 4 | 637 | |||
AY323979.1 | - | 5 | 725 | |||
BC014438.1 | - | 5 | 690 | 4877757 | ||
BC060853.1 | - | 5 | 709 | 5296086 |