Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284266.1 | NP_001271195.1 | 2, 3 | 287 | ||
NM_001284267.1 | NP_001271196.1 | 2, 3 | 140 | |||
NM_001284268.1 | NP_001271197.1 | 2, 3 | 298 | |||
NM_001284269.1 | NP_001271198.1 | 2, 3 | 298 | |||
NM_145231.3 | NP_660274.1 | 2, 3 | 287 | |||
GenBank mRNA | AK094740.1 | - | 2, 3 | 240 | ||
AK298930.1 | - | 2, 3 | 232 | |||
AL832321.1 | - | 2, 3 | 224 | |||
BC001787.1 | - | 2, 3 | 199 | 3353520 | ||
BG494945.1 | - | 2, 3 | 252 | |||
BP371056.1 | - | 2, 3 | 140 | |||
BX161414.1 | - | 2, 3 | 264 | |||
CF593330.1 | - | 2, 3 | 283 |