Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204426.1 | NP_001191355.1 | 7, 8 | 1065 | ||
NM_002314.3 | NP_002305.1 | 8, 9 | 1287 | |||
GenBank mRNA | AB209557.1 | - | 5, 6 | 759 | ||
AF134379.1 | - | 8, 9 | 1012 | |||
AK026544.1 | - | 2, 3 | 323 | |||
AK290162.1 | - | 9, 10 | 1212 | |||
AK300382.1 | - | 7, 8 | 1004 | |||
D26309.1 | - | 9, 10 | 1168 |