Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FANK1 SIRNA Assays ›
Gene Symbol:
FANK1
Gene Name:
fibronectin type III and ankyrin repeat domains 1
Gene Aliases:
HSD13
Chromosome Location:
Chr.10: 125896539 - 126009599 on Build GRCh38
UniGene ID:
Hs.352591
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
131800
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_145235.3
NP_660278.3
11
1175
XM_005270280.3
XP_005270337.1
10
991
XM_006718058.3
XP_006718121.1
13
1174
XM_006718064.3
XP_006718127.1
12
1168
XM_011540348.2
XP_011538650.1
13
1289
XM_011540349.2
XP_011538651.1
13
1293
XM_011540350.2
XP_011538652.1
12
1252
XM_011540351.2
XP_011538653.1
12
1228
XM_011540356.2
XP_011538658.1
11
1140
XM_011540357.2
XP_011538659.1
11
1113
XM_011540358.2
XP_011538660.1
9
1032
XM_011540360.2
XP_011538662.1
11
1069
XM_011540361.2
XP_011538663.1
11
1004
XM_011540362.2
XP_011538664.1
9
890
XM_017016916.1
XP_016872405.1
13
1235
XM_017016917.1
XP_016872406.1
13
1370
XM_017016918.1
XP_016872407.1
12
1177
XM_017016919.1
XP_016872408.1
12
1329
XM_017016920.1
XP_016872409.1
13
1386
XM_017016921.1
XP_016872410.1
12
1292
XM_017016922.1
XP_016872411.1
12
1192
XM_017016923.1
XP_016872412.1
9
1051
XM_017016924.1
XP_016872413.1
14
1347
XM_017016925.1
XP_016872414.1
8
7774
XM_017016926.1
XP_016872415.1
8
1304
GenBank mRNA
AK021437.1
-
3
1464
AY251163.1
-
9
1182
AY358154.1
-
12
1231
BC024189.2
-
11
1175
4826296
CR627249.1
-
8
7766
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-8skhh:80/10.239.164.194:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline