Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113525.1 | NP_001106997.1 | 2 | 494 | ||
NM_152287.3 | NP_689500.2 | 2 | 484 | |||
NR_110122.1 | 2 | 484 | ||||
NR_110126.1 | 2 | 484 | ||||
NR_110128.1 | 2 | 407 | ||||
NR_110129.1 | 2 | 496 | ||||
XM_005256324.3 | XP_005256381.1 | 2 | 494 | |||
XM_017023889.1 | XP_016879378.1 | 2 | 494 | |||
XM_017023890.1 | XP_016879379.1 | 1 | 451 | |||
GenBank mRNA | AK026482.1 | - | 2 | 466 | ||
AK056405.1 | - | 2 | 435 | |||
AK289801.1 | - | 2 | 375 | |||
BC032781.1 | - | 1 | 153 | |||
BC105959.1 | - | 2 | 309 | 40036181 | ||
BC107603.1 | - | 2 | 309 | 40036180 | ||
BC110348.1 | - | 2 | 209 | 5196957 | ||
BM921035.1 | - | 2 | 465 | |||
DA227038.1 | - | 2 | 484 | |||
DA523531.1 | - | 2 | 407 |