Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044385.2 | NP_001037850.1 | 9 | 899 | ||
NM_152388.3 | NP_689601.2 | 9 | 1064 | |||
GenBank mRNA | AB053301.1 | - | 9 | 1066 | ||
AF212043.1 | - | 4 | 468 | |||
AK090601.1 | - | 9 | 757 | |||
AK303954.1 | - | 9 | 886 | |||
AK304395.1 | - | 7 | 588 | |||
BC029611.1 | - | 9 | 812 | 5304046 | ||
BQ219059.1 | - | 9 | 872 |