Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFAND4 SIRNA Assays ›
Gene Symbol:
ZFAND4
Gene Name:
zinc finger AN1-type containing 4
Gene Aliases:
ANUBL1
Chromosome Location:
Chr.10: 45614567 - 45672962 on Build GRCh38
UniGene ID:
Hs.659717
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
131852
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFAND4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128324.2
NP_001121796.1
8, 9
2182
NM_001282905.1
NP_001269834.1
9, 10
2632
NM_001282906.1
NP_001269835.1
9, 10
2284
NM_174890.3
NP_777550.2
8, 9
2530
XM_005271842.2
XP_005271899.1
7, 8
2055
XM_005271843.2
XP_005271900.1
7, 8
2222
XM_005271844.3
XP_005271901.1
5, 6
1688
XM_011540366.2
XP_011538668.1
6, 7
2126
XM_011540368.1
XP_011538670.1
5, 6
1571
XM_017016931.1
XP_016872420.1
6, 7
2146
XM_017016932.1
XP_016872421.1
8, 9
2292
XM_017016933.1
XP_016872422.1
8, 9
2159
XM_017016934.1
XP_016872423.1
8, 9
2212
XM_017016936.1
XP_016872425.1
6, 7
2302
XM_017016937.1
XP_016872426.1
6, 7
2150
XM_017016938.1
XP_016872427.1
6, 7
2444
XM_017016939.1
XP_016872428.1
6, 7
2015
XM_017016940.1
XP_016872429.1
9, 10
2708
XM_017016941.1
XP_016872430.1
9, 10
2251
XM_017016942.1
XP_016872431.1
7, 8
2240
XM_017016943.1
XP_016872432.1
7, 8
2516
XM_017016944.1
XP_016872433.1
7, 8
2073
XM_017016945.1
XP_016872434.1
5, 6
1660
XM_017016946.1
XP_016872435.1
9, 10
2326
GenBank mRNA
AF311324.1
-
8, 9
2146
AK097504.1
-
5, 6
1650
AK292420.1
-
9, 10
2233
AK292469.1
-
8, 9
2131
AK314395.1
-
8, 9
2142
BC045587.1
-
8, 9
2182
5269483
BC048968.1
-
9, 10
2174
5272133
Back To Top
More Information
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-g5l8q:80/100.66.79.176:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline