Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKHD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177531.4 | NP_803875.2 | 37 | 4808 | ||
XM_011517371.2 | XP_011515673.1 | 37 | 4759 | |||
XM_017013969.1 | XP_016869458.1 | 37 | 4759 | |||
XM_017013970.1 | XP_016869459.1 | 36 | 4645 | |||
XM_017013971.1 | XP_016869460.1 | 37 | 4759 | |||
XM_017013972.1 | XP_016869461.1 | 35 | 4456 | |||
XM_017013973.1 | XP_016869462.1 | 13 | 1815 | |||
XM_017013974.1 | XP_016869463.1 | 8 | 1315 | |||
GenBank mRNA | AY219181.1 | - | 37 | 4808 |