Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182640.2 | NP_872578.1 | 6, 7 | 648 | ||
XM_011511644.2 | XP_011509946.1 | 5, 6 | 488 | |||
GenBank mRNA | AK130078.1 | - | 6, 7 | 589 | ||
BC036575.1 | - | 7, 8 | 601 | |||
BC047784.1 | - | 5, 6 | 298 | |||
BC057240.1 | - | 6, 7 | 602 | 6495350 | ||
DT217261.1 | - | 6, 7 | 648 |