Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTAT33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI381531.1 | - | 1 | 212 |
Gene Symbol: MRPS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182640.2 | NP_872578.1 | 10 | 1103 | ||
XM_011511644.2 | XP_011509946.1 | 9 | 943 | |||
GenBank mRNA | AK001451.1 | - | 7 | 636 | ||
AK130078.1 | - | 10 | 1044 | |||
BC036575.1 | - | 11 | 1056 | |||
BC047784.1 | - | 9 | 753 | |||
BC057240.1 | - | 10 | 1057 | 6495350 |