Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182640.2 | NP_872578.1 | 10, 11 | 1175 | ||
XM_011511644.2 | XP_011509946.1 | 9, 10 | 1015 | |||
GenBank mRNA | AK001451.1 | - | 7, 8 | 708 | ||
AK130078.1 | - | 10, 11 | 1116 | |||
BC036575.1 | - | 11, 12 | 1128 | |||
BC047784.1 | - | 9, 10 | 825 | |||
BC057240.1 | - | 10, 11 | 1129 | 6495350 |