Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033058.2 | NP_149047.2 | 11 | 2648 | ||
NM_184085.1 | NP_908973.1 | 10 | 2560 | |||
NM_184086.1 | NP_908974.1 | 9 | 2272 | |||
NM_184087.1 | NP_908975.1 | 5 | 1639 | |||
XM_005251316.4 | XP_005251373.1 | 12 | 3582 | |||
XM_011517613.2 | XP_011515915.1 | 12 | 3585 | |||
XM_011517614.2 | XP_011515916.1 | 12 | 3508 | |||
XM_011517615.2 | XP_011515917.1 | 13 | 3673 | |||
XM_011517616.2 | XP_011515918.1 | 11 | 3297 | |||
XM_011517617.2 | XP_011515919.1 | 10 | 2642 | |||
XM_017013908.1 | XP_016869397.1 | 10 | 2278 | |||
XM_017013909.1 | XP_016869398.1 | 9 | 2097 | |||
GenBank mRNA | AK091310.1 | - | 9 | 2422 |