Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318716.1 | NP_001305645.1 | 4 | 1781 | ||
NM_001318717.1 | NP_001305646.1 | 4 | 1702 | |||
NM_001318718.1 | NP_001305647.1 | 4 | 1682 | |||
NM_001318719.1 | NP_001305648.1 | 4 | 1660 | |||
NM_001318720.1 | NP_001305649.1 | 4 | 1654 | |||
NM_001318721.1 | NP_001305650.1 | 4 | 1629 | |||
NM_001318722.1 | NP_001305651.1 | 4 | 1650 | |||
NM_001318723.1 | NP_001305652.1 | 4 | 1734 | |||
NM_001318724.1 | NP_001305653.1 | 4 | 1734 | |||
NM_001318725.1 | NP_001305654.1 | 4 | 1637 | |||
NM_016506.6 | NP_057590.3 | 4 | 1738 | |||
NM_018095.5 | NP_060565.4 | 4 | 1633 | |||
XM_017018009.1 | XP_016873498.1 | 4 | 1617 | |||
GenBank mRNA | AK001312.1 | - | 4 | 1587 | ||
AK001749.1 | - | 4 | 1674 | |||
AK091607.1 | - | 3 | 2311 | |||
AK222550.1 | - | 4 | 1584 | |||
AK225062.1 | - | 4 | 1584 | |||
AL713735.1 | - | 3 | 1563 | |||
BC002736.2 | - | 4 | 1675 | 3628290 | ||
CR457270.1 | - | 3 | 1521 |