Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017415.1 | NP_001017415.1 | 4 | 689 | ||
NM_001017416.1 | NP_001017416.1 | 4 | 594 | |||
NM_003368.4 | NP_003359.3 | 4 | 1119 | |||
GenBank mRNA | AB014458.1 | - | 4 | 470 | ||
AB208893.1 | - | 4 | 751 | |||
AF117386.1 | - | 4 | 606 | |||
AK312423.1 | - | 4 | 782 | |||
AL117575.1 | - | 3 | 441 | |||
BC018745.1 | - | 4 | 1119 | |||
BC032364.1 | - | 4 | 769 | |||
BC050525.1 | - | 4 | 823 | 6473568 |