Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322270.1 | NP_001309199.1 | 15 | 3138 | ||
NM_001322271.1 | NP_001309200.1 | 15 | 3131 | |||
NM_001322272.1 | NP_001309201.1 | 16 | 3496 | |||
NM_017437.2 | NP_059133.1 | 16 | 3291 | |||
GenBank mRNA | AB037788.1 | - | 10 | 2415 | ||
AK023583.1 | - | 13 | 2870 | |||
AL442079.1 | - | 9 | 2191 |