Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVRAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003369.3 | NP_003360.2 | 7 | 879 | ||
XM_011545226.2 | XP_011543528.1 | 7 | 1194 | |||
XM_011545227.2 | XP_011543529.1 | 7 | 1194 | |||
XM_011545228.2 | XP_011543530.1 | 7 | 1194 | |||
XM_017018224.1 | XP_016873713.1 | 5 | 633 | |||
XM_017018225.1 | XP_016873714.1 | 5 | 633 | |||
XM_017018226.1 | XP_016873715.1 | 7 | 1194 | |||
XM_017018227.1 | XP_016873716.1 | 1 | 45 | |||
GenBank mRNA | AB012958.1 | - | 7 | 816 | ||
AK222772.1 | - | 7 | 770 | |||
AK291011.1 | - | 7 | 857 | |||
AK293782.1 | - | 7 | 620 | |||
AK311341.1 | - | 2 | 98 | |||
BC064837.1 | - | 7 | 789 | 6186851 |