Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017539.2 | NP_060009.1 | 2 | 213 | ||
XM_006721062.1 | XP_006721125.1 | 3 | 402 | |||
XM_011545882.2 | XP_011544184.1 | 4 | 842 | |||
XM_011545883.1 | XP_011544185.1 | 4 | 610 | |||
XM_011545885.2 | XP_011544187.1 | 4 | 842 | |||
XM_017023426.1 | XP_016878915.1 | 4 | 842 | |||
XM_017023427.1 | XP_016878916.1 | 4 | 842 | |||
XM_017023428.1 | XP_016878917.1 | 4 | 842 | |||
XM_017023429.1 | XP_016878918.1 | 4 | 843 | |||
XM_017023431.1 | XP_016878920.1 | 4 | 843 | |||
XM_017023432.1 | XP_016878921.1 | 2 | 309 | |||
GenBank mRNA | AF494040.1 | - | 2 | 213 | ||
AK056509.2 | - | 3 | 402 | |||
AK303958.1 | - | 3 | 411 | |||
AK310973.1 | - | 3 | 399 |