Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017539.2 | NP_060009.1 | 10, 11 | 1592 | ||
XM_017023432.1 | XP_016878921.1 | 10, 11 | 1688 | |||
GenBank mRNA | AF494040.1 | - | 10, 11 | 1592 |