Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314014.1 | NP_001300943.1 | 3 | 384 | ||
NM_017542.4 | NP_060012.3 | 5 | 610 | |||
XM_005245370.3 | XP_005245427.1 | 5 | 731 | |||
XM_011509814.2 | XP_011508116.1 | 5 | 887 | |||
XM_011509815.1 | XP_011508117.1 | 5 | 542 | |||
XM_017001914.1 | XP_016857403.1 | 5 | 604 | |||
XM_017001915.1 | XP_016857404.1 | 3 | 272 | |||
GenBank mRNA | AB040946.1 | - | 5 | 527 | ||
AK056711.1 | - | 5 | 508 | |||
AK293946.1 | - | 3 | 344 | |||
AK299537.1 | - | 4 | 443 | |||
BC131624.1 | - | 5 | 495 | 40117695 |