Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318864.1 | NP_001305793.1 | 14 | 2506 | ||
NM_017563.4 | NP_060033.3 | 13 | 2497 | |||
XM_005265238.4 | XP_005265295.1 | 13 | 2708 | |||
XM_006713209.3 | XP_006713272.1 | 15 | 2635 | |||
XM_011533849.2 | XP_011532151.2 | 13 | 2292 | |||
GenBank mRNA | AF458067.1 | - | 13 | 2384 | ||
AF494208.1 | - | 13 | 2473 | |||
AF494211.1 | - | 14 | 2474 | |||
AK001251.1 | - | 1 | 41 | |||
AK093074.1 | - | 14 | 2473 | |||
AL133097.1 | - | 9 | 1859 | |||
AL833913.1 | - | 13 | 2297 | |||
BC038369.1 | - | 14 | 2496 | |||
BC111702.1 | - | 13 | 2324 | 40038598 |