Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317774.1 | NP_001304703.1 | 2, 3 | 381 | ||
NM_017578.3 | NP_060048.2 | 3, 4 | 461 | |||
XM_011512933.2 | XP_011511235.1 | 3, 4 | 425 | |||
XM_011512934.2 | XP_011511236.1 | 3, 4 | 536 | |||
GenBank mRNA | AF303889.2 | - | 3, 4 | 461 | ||
BC132744.1 | - | 1, 2 | 217 | 40146766 | ||
BC132746.1 | - | 1, 2 | 218 | 40146768 | ||
BG724071.1 | - | 2, 3 | 349 |