Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARMIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173977.1 | NP_001167448.1 | 33 | 3740 | ||
NM_017640.5 | NP_060110.4 | 33 | 3740 | |||
XM_005249218.1 | XP_005249275.1 | 33 | 3740 | |||
XM_017011008.1 | XP_016866497.1 | 35 | 3770 | |||
XM_017011009.1 | XP_016866498.1 | 35 | 3770 | |||
XM_017011010.1 | XP_016866499.1 | 34 | 3758 | |||
XM_017011011.1 | XP_016866500.1 | 34 | 3752 | |||
XM_017011012.1 | XP_016866501.1 | 35 | 3770 | |||
XM_017011013.1 | XP_016866502.1 | 34 | 3752 | |||
XM_017011014.1 | XP_016866503.1 | 34 | 3710 | |||
XM_017011015.1 | XP_016866504.1 | 35 | 3770 | |||
XM_017011016.1 | XP_016866505.1 | 34 | 3752 | |||
GenBank mRNA | AK000055.1 | - | 8 | 1077 | ||
AK225208.1 | - | 8 | 1077 | |||
BC140770.1 | - | 33 | 3521 | 9021638 | ||
FJ009082.1 | - | 33 | 3372 |