Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270890.1 | NP_001257819.1 | 2 | 443 | ||
NM_017645.4 | NP_060115.3 | 2 | 443 | |||
GenBank mRNA | AF533709.1 | - | 2 | 625 | ||
AF537091.1 | - | 2 | 405 | |||
AK056458.1 | - | 2 | 405 | |||
AL832495.1 | - | 2 | 403 | |||
BC026178.1 | - | 2 | 406 | |||
BC113981.1 | - | 2 | 240 | 40082296 | ||
BC114492.1 | - | 2 | 240 | 40082291 | ||
DB010660.1 | - | 2 | 443 |