Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270890.1 | NP_001257819.1 | 6 | 896 | ||
NM_017645.4 | NP_060115.3 | 6 | 896 | |||
XM_017014830.1 | XP_016870319.1 | 2 | 47 | |||
GenBank mRNA | AF533709.1 | - | 6 | 1078 | ||
AF537091.1 | - | 6 | 858 | |||
AK022964.1 | - | 3 | 212 | |||
AK056458.1 | - | 6 | 858 | |||
AL832495.1 | - | 6 | 856 | |||
BC026178.1 | - | 6 | 859 | |||
BC065935.1 | - | 4 | 396 | |||
BC111041.1 | - | 2 | 172 | 5529130 | ||
BC113981.1 | - | 6 | 693 | 40082296 | ||
BC114492.1 | - | 6 | 693 | 40082291 |