Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270890.1 | NP_001257819.1 | 8 | 1132 | ||
NM_017645.4 | NP_060115.3 | 8 | 1132 | |||
XM_017014830.1 | XP_016870319.1 | 4 | 283 | |||
GenBank mRNA | AF533709.1 | - | 8 | 1314 | ||
AF537091.1 | - | 8 | 1094 | |||
AK022964.1 | - | 5 | 448 | |||
AK023068.1 | - | 1 | 51 | |||
AK056458.1 | - | 8 | 1094 | |||
AL832495.1 | - | 8 | 1092 | |||
BC026178.1 | - | 8 | 1095 | |||
BC065935.1 | - | 6 | 632 | |||
BC071625.1 | - | 1 | 91 | |||
BC111041.1 | - | 4 | 408 | 5529130 | ||
BC113981.1 | - | 8 | 929 | 40082296 | ||
BC114492.1 | - | 8 | 929 | 40082291 |