Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134830.1 | NP_001128302.1 | 6 | 1120 | ||
NM_001134831.1 | NP_001128303.1 | 8 | 1267 | |||
NM_001134832.1 | NP_001128304.1 | 7 | 1182 | |||
NM_017651.4 | NP_060121.3 | 7 | 1182 | |||
XM_011535910.2 | XP_011534212.1 | 9 | 1411 | |||
XM_011535911.2 | XP_011534213.1 | 7 | 1141 | |||
XM_017010977.1 | XP_016866466.1 | 8 | 1386 | |||
XM_017010978.1 | XP_016866467.1 | 7 | 1084 | |||
XM_017010979.1 | XP_016866468.1 | 7 | 1084 | |||
XM_017010980.1 | XP_016866469.1 | 7 | 1084 | |||
XM_017010981.1 | XP_016866470.1 | 7 | 1219 | |||
XM_017010982.1 | XP_016866471.1 | 7 | 1084 | |||
XM_017010983.1 | XP_016866472.1 | 7 | 1084 | |||
XM_017010984.1 | XP_016866473.1 | 7 | 1084 | |||
GenBank mRNA | AJ459824.1 | - | 7 | 1162 | ||
AJ459825.1 | - | 7 | 1162 | |||
AJ606362.1 | - | 6 | 1100 | |||
AK092262.1 | - | 6 | 1093 | |||
AL136797.1 | - | 5 | 901 | |||
BC029417.2 | - | 7 | 1083 | |||
BC065712.1 | - | 5 | 946 | |||
BC094800.1 | - | 7 | 1157 | 4823262 | ||
DQ090887.1 | - | 8 | 1221 |