Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EZR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111077.1 | NP_001104547.1 | 14 | 2529 | ||
NM_003379.4 | NP_003370.2 | 13 | 2563 | |||
XM_011536110.1 | XP_011534412.1 | 10 | 1940 | |||
GenBank mRNA | AK129727.1 | - | 7 | 1485 | ||
AK316031.1 | - | 4 | 1286 | |||
BC013903.2 | - | 13 | 2480 | 2959399 | ||
BC068458.1 | - | 13 | 2489 | 4819793 | ||
J05021.1 | - | 13 | 2422 |