Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193335.1 | NP_001180264.1 | 4 | 716 | ||
NM_017680.4 | NP_060150.4 | 4 | 716 | |||
GenBank mRNA | AF316824.1 | - | 3 | 434 | ||
AK000136.1 | - | 2 | 86 | |||
AK027359.1 | - | 4 | 519 | |||
AK304501.1 | - | 4 | 523 | |||
AK308107.1 | - | 4 | 630 | |||
AY029191.1 | - | 4 | 649 | |||
AY358329.1 | - | 4 | 649 | |||
BC063114.1 | - | 4 | 681 | 30404602 |