Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017686.3 | NP_060156.1 | 14 | 2200 | ||
XM_005270959.2 | XP_005271016.1 | 15 | 2266 | |||
XM_011541623.2 | XP_011539925.1 | 15 | 2296 | |||
XM_017001531.1 | XP_016857020.1 | 16 | 2288 | |||
XM_017001532.1 | XP_016857021.1 | 15 | 2643 | |||
GenBank mRNA | AK000149.1 | - | 14 | 2103 |