Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316936.1 | NP_001303865.1 | 2 | 1593 | ||
NM_001316937.1 | NP_001303866.1 | 3 | 1666 | |||
NM_001316938.1 | NP_001303867.1 | 4 | 1736 | |||
NM_001316939.1 | NP_001303868.1 | 4 | 1710 | |||
NM_001316940.1 | NP_001303869.1 | 3 | 1588 | |||
NM_001316941.1 | NP_001303870.1 | 2 | 1619 | |||
NM_001316942.1 | NP_001303871.1 | 4 | 1764 | |||
NM_017703.2 | NP_060173.1 | 3 | 1692 | |||
XM_006722782.3 | XP_006722845.2 | 2 | 1443 | |||
XM_017026912.1 | XP_016882401.1 | 3 | 1568 | |||
GenBank mRNA | AK000195.1 | - | 3 | 1644 | ||
AK027004.1 | - | 4 | 1711 | |||
AK093760.1 | - | 3 | 1639 | |||
BC001586.2 | - | 3 | 1596 | 2966439 | ||
BX647073.1 | - | 2 | 1529 |