Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PXK SIRNA Assays ›
Gene Symbol:
PXK
Gene Name:
PX domain containing serine/threonine kinase like
Gene Aliases:
MONaKA
Chromosome Location:
Chr.3: 58332868 - 58426127 on Build GRCh38
UniGene ID:
Hs.190544
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
132074
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PXK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289095.1
NP_001276024.1
7
705
NM_001289096.1
NP_001276025.1
6
657
NM_001289098.1
NP_001276027.1
8
756
NM_001289099.1
NP_001276028.1
8
861
NM_001289100.1
NP_001276029.1
6
657
NM_001289101.1
NP_001276030.1
5
470
NM_017771.4
NP_060241.2
8
756
XM_005265250.4
XP_005265307.1
8
756
XM_005265252.4
XP_005265309.1
8
756
XM_005265255.3
XP_005265312.1
8
756
XM_005265256.3
XP_005265313.1
8
756
XM_017006671.1
XP_016862160.1
7
647
XM_017006672.1
XP_016862161.1
7
647
XM_017006673.1
XP_016862162.1
6
648
XM_017006674.1
XP_016862163.1
7
647
XM_017006675.1
XP_016862164.1
8
756
XM_017006676.1
XP_016862165.1
8
756
XM_017006677.1
XP_016862166.1
8
756
XM_017006678.1
XP_016862167.1
8
756
XM_017006679.1
XP_016862168.1
8
756
XM_017006680.1
XP_016862169.1
8
756
XM_017006681.1
XP_016862170.1
7
647
XM_017006682.1
XP_016862171.1
7
647
XM_017006683.1
XP_016862172.1
7
647
XM_017006684.1
XP_016862173.1
7
647
XM_017006685.1
XP_016862174.1
7
647
XM_017006686.1
XP_016862175.1
9
870
XM_017006687.1
XP_016862176.1
9
868
XM_017006688.1
XP_016862177.1
6
632
XM_017006689.1
XP_016862178.1
6
622
XM_017006690.1
XP_016862179.1
6
613
XM_017006691.1
XP_016862180.1
5
469
XM_017006692.1
XP_016862181.1
9
907
XM_017006693.1
XP_016862182.1
9
907
XM_017006694.1
XP_016862183.1
9
801
XM_017006695.1
XP_016862184.1
9
907
XM_017006696.1
XP_016862185.1
5
469
XM_017006697.1
XP_016862186.1
5
464
XM_017006698.1
XP_016862187.1
5
462
XM_017006699.1
XP_016862188.1
5
474
XM_017006700.1
XP_016862189.1
5
492
XM_017006701.1
XP_016862190.1
5
469
XM_017006702.1
XP_016862191.1
5
469
XM_017006703.1
XP_016862192.1
5
469
XM_017006704.1
XP_016862193.1
5
469
GenBank mRNA
AB209688.1
-
8
734
AF399753.1
-
8
657
AK000342.1
-
8
754
AK131385.1
-
7
647
AK295542.1
-
6
649
AK296034.1
-
5
328
AK296215.1
-
5
405
AY274811.1
-
8
756
AY437879.1
-
6
600
AY847220.1
-
8
764
AY847221.1
-
5
373
AY847222.1
-
5
432
BC008943.2
-
8
737
BC014479.1
-
8
672
4856139
DQ124707.1
-
8
657
DQ124708.1
-
8
657
KF774202.1
-
8
766
KF774203.1
-
8
766
KF774204.1
-
8
841
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
inflammatory response
negative regulation of ATPase activity
regulation of membrane potential
negative regulation of ion transport
modulation of synaptic transmission
nucleotide binding
actin binding
protein kinase activity
ATP binding
protein C-terminus binding
phosphatidylinositol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline