Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D22B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017772.3 | NP_060242.2 | 2 | 318 | ||
NR_130108.1 | 2 | 318 | ||||
XM_011514738.2 | XP_011513040.1 | 2 | 270 | |||
XM_011514739.1 | XP_011513041.1 | 2 | 270 | |||
GenBank mRNA | AB449909.1 | - | 2 | 104 | ||
AK096340.1 | - | 2 | 248 | |||
AK292893.1 | - | 2 | 213 | |||
AY313781.1 | - | 2 | 250 | |||
BC047658.1 | - | 2 | 250 | |||
BC109026.1 | - | 2 | 248 | 40027698 | ||
BC109027.1 | - | 2 | 248 | 40027700 | ||
BX648317.1 | - | 2 | 249 | |||
DA726945.1 | - | 2 | 318 |