Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOCAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017794.4 | NP_060264.4 | 10 | 1098 | ||
XM_005251494.3 | XP_005251551.2 | 8 | 1082 | |||
XM_011517945.2 | XP_011516247.1 | 7 | 686 | |||
XM_017014852.1 | XP_016870341.1 | 9 | 872 | |||
XM_017014853.1 | XP_016870342.1 | 9 | 872 | |||
XM_017014854.1 | XP_016870343.1 | 8 | 767 | |||
XM_017014855.1 | XP_016870344.1 | 9 | 971 | |||
XM_017014856.1 | XP_016870345.1 | 9 | 912 | |||
XM_017014857.1 | XP_016870346.1 | 8 | 1082 | |||
XM_017014858.1 | XP_016870347.1 | 7 | 977 | |||
XM_017014859.1 | XP_016870348.1 | 4 | 518 | |||
GenBank mRNA | AK056522.1 | - | 10 | 1098 | ||
AY139834.1 | - | 10 | 1097 |