Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOCAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017794.4 | NP_060264.4 | 16 | 2137 | ||
XM_005251494.3 | XP_005251551.2 | 14 | 2121 | |||
XM_011517945.2 | XP_011516247.1 | 13 | 1725 | |||
XM_017014852.1 | XP_016870341.1 | 15 | 1911 | |||
XM_017014853.1 | XP_016870342.1 | 14 | 1806 | |||
XM_017014854.1 | XP_016870343.1 | 14 | 1806 | |||
XM_017014855.1 | XP_016870344.1 | 15 | 2010 | |||
XM_017014856.1 | XP_016870345.1 | 15 | 1951 | |||
XM_017014857.1 | XP_016870346.1 | 13 | 2016 | |||
XM_017014858.1 | XP_016870347.1 | 13 | 2016 | |||
XM_017014859.1 | XP_016870348.1 | 10 | 1557 | |||
GenBank mRNA | AB058700.1 | - | 2 | 243 | ||
AK056522.1 | - | 16 | 2137 | |||
AY139834.1 | - | 16 | 2136 | |||
DB288580.1 | - | 4 | 525 |