Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039140.2 | NP_001034229.1 | 6 | 1336 | ||
NM_001258429.1 | NP_001245358.1 | 6 | 1261 | |||
NM_001258430.1 | NP_001245359.1 | 6 | 1715 | |||
NR_047675.1 | 6 | 1405 | ||||
XM_011529266.2 | XP_011527568.1 | 5 | 1193 | |||
GenBank mRNA | AK000557.1 | - | 6 | 1200 | ||
AK024220.1 | - | 6 | 1200 | |||
AK097645.1 | - | 6 | 1843 | |||
AK124341.1 | - | 4 | 2453 | |||
AK125202.1 | - | 3 | 2407 | |||
AK225109.1 | - | 6 | 1200 | |||
AK290955.1 | - | 6 | 1405 | |||
BC010873.1 | - | 1 | 266 | |||
BC012196.1 | - | 6 | 1187 | 2822218 | ||
BC024036.1 | - | 6 | 1198 | 3510659 |