Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017882.2 | NP_060352.1 | 4 | 504 | ||
GenBank mRNA | AK000568.1 | - | 4 | 449 | ||
AK027604.1 | - | 4 | 472 | |||
AK291175.1 | - | 4 | 431 | |||
AK293197.1 | - | 4 | 680 | |||
BC010849.1 | - | 4 | 418 | 3878776 | ||
BC013130.1 | - | 4 | 355 | 4517748 | ||
CR457244.1 | - | 4 | 346 | |||
KU178671.1 | - | 4 | 346 | |||
KU178672.1 | - | 4 | 346 | |||
KU178673.1 | - | 3 | 247 |