Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017895.7 | NP_060365.7 | 10 | 1227 | ||
XM_011528888.2 | XP_011527190.1 | 11 | 1291 | |||
GenBank mRNA | AF193054.1 | - | 7 | 785 | ||
AF336851.1 | - | 4 | 461 | |||
AK022979.1 | - | 10 | 1097 | |||
AK025891.1 | - | 6 | 592 | |||
AK300135.1 | - | 10 | 1106 | |||
AL832131.1 | - | 10 | 1143 | |||
AY044431.1 | - | 10 | 1166 | |||
BC009304.1 | - | 10 | 1085 | |||
BC011927.2 | - | 10 | 1081 | |||
BC016060.1 | - | 10 | 1081 | |||
BC126287.1 | - | 10 | 1216 | 8992003 | ||
BC130275.1 | - | 10 | 1216 | 40146306 | ||
BC144125.1 | - | 10 | 1216 | 9052645 | ||
EU718675.1 | - | 10 | 1134 |