Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017895.7 | NP_060365.7 | 18 | 2206 | ||
XM_011528888.2 | XP_011527190.1 | 19 | 2270 | |||
GenBank mRNA | AF336851.1 | - | 12 | 1440 | ||
AK000603.1 | - | 9 | 981 | |||
AK022979.1 | - | 18 | 2076 | |||
AK025891.1 | - | 14 | 1571 | |||
AK127128.1 | - | 5 | 3038 | |||
AK223116.1 | - | 9 | 981 | |||
AK300135.1 | - | 18 | 2085 | |||
AL512707.1 | - | 5 | 434 | |||
AL832131.1 | - | 16 | 4410 | |||
AY044431.1 | - | 18 | 2145 | |||
BC009304.1 | - | 18 | 2064 | |||
BC011927.2 | - | 18 | 2060 | |||
BC016060.1 | - | 18 | 2060 | |||
BC126287.1 | - | 18 | 2195 | 8992003 | ||
BC130275.1 | - | 18 | 2195 | 40146306 | ||
BC144125.1 | - | 18 | 2195 | 9052645 | ||
EU718675.1 | - | 18 | 2113 |
Gene Symbol: SS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF161377.1 | - | 3 | 376 |