Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAK1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017906.2 | NP_060376.2 | 1, 2 | 123 | ||
GenBank mRNA | AF283303.1 | - | 1, 2 | 166 | ||
AK000631.1 | - | 1, 2 | 171 | |||
AK225516.1 | - | 1, 2 | 171 | |||
BC010907.1 | - | 1, 2 | 127 | 4251347 | ||
KU178675.1 | - | 1, 2 | 92 |