Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017924.3 | NP_060394.1 | 2 | 479 | ||
XM_017021390.1 | XP_016876879.1 | 2 | 653 | |||
GenBank mRNA | AF061731.1 | - | 2 | 449 | ||
AK000678.1 | - | 2 | 404 | |||
BC009645.1 | - | 2 | 374 | 3458006 | ||
BC056255.1 | - | 2 | 450 | 4694305 | ||
BC061637.1 | - | 2 | 414 | 5532778 | ||
CR457248.1 | - | 1 | 332 | |||
DA605680.1 | - | 2 | 479 |