Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256394.1 | NP_001243323.1 | 2, 3 | 457 | ||
NM_017948.5 | NP_060418.4 | 2, 3 | 469 | |||
NR_046106.1 | 2, 3 | 469 | ||||
XM_006717166.3 | XP_006717229.1 | 2, 3 | 335 | |||
XM_006717167.3 | XP_006717230.1 | 2, 3 | 457 | |||
XM_006717168.3 | XP_006717231.1 | 2, 3 | 228 | |||
XM_006717169.3 | XP_006717232.1 | 2, 3 | 372 | |||
XM_006717170.3 | XP_006717233.1 | 2, 3 | 240 | |||
XM_006717172.3 | XP_006717235.1 | 2, 3 | 240 | |||
XM_006717173.3 | XP_006717236.1 | 2, 3 | 240 | |||
XM_017014876.1 | XP_016870365.1 | 2, 3 | 335 | |||
XM_017014877.1 | XP_016870366.1 | 2, 3 | 240 | |||
XM_017014878.1 | XP_016870367.1 | 2, 3 | 366 | |||
XM_017014879.1 | XP_016870368.1 | 2, 3 | 198 | |||
XM_017014880.1 | XP_016870369.1 | 2, 3 | 240 | |||
GenBank mRNA | AB105104.1 | - | 2, 3 | 435 | ||
AB109030.1 | - | 1, 2 | 132 | |||
AK001049.1 | - | 2, 3 | 351 | |||
AK297355.1 | - | 2, 3 | 352 | |||
AK307408.1 | - | 2, 3 | 360 | |||
BC146810.1 | - | 2, 3 | 213 | 8860369 | ||
CR627363.1 | - | 2, 3 | 213 | |||
DA470513.1 | - | 2, 3 | 459 |